Water Agar, HiMedia Laboratories
Supplier: HiMedia
M1366-500G M1366-2.5KG M1366-5KG
95021-024EA 195.06 USD
95021-024 89223-868 89223-870
Water Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Water Agar is recommended for enumeration, cultivation and observation of sporulation of some fungi.
  • Appearance
  • : White to cream homogeneous free flowing powder
  • Gelling: Firm, comparable with 2.0% agar gel
  • Colour and clarity of prepared medium: White coloured clear to slightly opalescent gel forms in petri plates
Water Agar is often recommended for enumeration, cultivation and observation of sporulation of some fungi
Water Agar is used for enumeration of fungi according to MPN method.
Direct Plating is considered to be one of the most effective techniques for mycological examination of all foods.