WHEATON® MicroLiter Snap-Top Vials, Component Kits, Unassembled, 11 mm, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
11-5051G 11-5051K 11-5053DB 11-5053K 11-5054DB 11-5253K 11-5253N 11-5254G 11-5254N 11-5255N 11-5255R 11-6255R 11-5351G-500 11-5351R-500 11-5353DB-500 11-5353K-700 11-5353N-700 11-5354DB-500 11-5354G-700 11-5355B-700 11-5355K-100
97047-366PK 61.28 USD
97047-366 97047-368 97047-370 97047-372 97047-374 97047-378 97047-380 97047-382 97047-384 97047-386 97047-388 97047-392 97047-394 97047-396 97047-398 97047-400 97047-402 97047-406 97047-408 97047-410 97047-412 97047-414 97047-416 97047-420 97047-422 97047-424 97047-426 97047-428 97047-430 97047-434 97047-444 97047-446 97047-448 97047-450 97047-452 97047-456 97047-458 97047-460 97047-462 97047-464 97047-466 97047-470 97047-472 97047-474 97047-476 97047-478 97047-480 97047-484 97047-486 97047-488 97047-490 97047-492 97047-494 97047-498 97047-500 97047-502 97047-504 97047-506 97047-508 97047-512 97047-526 97047-528 97047-530 97047-532 97047-534 97047-544 97047-546 97047-548 97047-550 97047-552 97047-556 97047-558 97047-560 97047-562 97047-564 97047-566 97047-570 97047-572 97047-574 97047-576 97047-578 97047-580 97047-584 97047-586 97047-588 97047-590 97047-592 97047-594 97047-598 97047-600 97047-602 97047-604 97047-606 97047-608 97047-612 97050-418 97050-419 97050-420 97050-421 97050-422 97050-423 97050-424 97050-425 97050-426 97050-427 97050-428 97050-429 97050-430 97050-431 97050-432 97050-433 97050-434 97050-435 97050-436 97050-437 97050-438 97050-439 97050-440 97050-441 97050-442 97050-443 97050-444 97050-445 97050-446 97050-447 97050-448 97050-449 97050-450 97050-451 97050-452 97050-453 97050-454 97050-455 97050-456 97050-457 97050-458 97050-459 97050-460 97050-461 97050-462 97050-463 97050-464 97050-465 97050-466 97050-467 97050-468 97050-469 97050-470 97050-471 97050-472 97050-473 97050-474 97050-475 97050-476 97050-477 97050-478 97050-479 97050-480 97050-481 97050-482 97050-483 97050-484 97050-485 97050-486 97050-487 97050-488 97050-489 97050-490 97050-491 97050-492 97050-493 97050-494 97050-495 97050-496 97050-497 97050-498 97050-499 97050-500 97050-501 97050-502
WHEATON® MicroLiter Snap-Top Vials, Component Kits, Unassembled, 11 mm, DWK Life Sciences
Vials
MicroLiter vials are identical, clean, dependable vials, certified to ensure optimal performance and reproducible results.
  • Unassembled kits
  • Multiple configurations available
MicroLiter vials are cleaned and packaged in a certified Class 10000 clean room according to a designed process, developed to eliminate chromatographic errors associated with vials.
Vials and inserts are made from Type 1 borosilicate glass or polypropylene and meet strict tolerances. They perform equal to an 11 mm crimp-top vial for samples that have low volatility, while offering the convenience and accessibility of snap caps. Vials are available in unassembled kits with blue polypropylene caps.
Critical physical dimensions are fully tested, as well as chemical cleanliness testing to ensure that trace levels of residues are not present.