γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥99%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 3-Morpholinopropane-1-sulfonic acid, 3-(N-Morpholino)propanesulfonic acid, 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid

0219483725 0219483780 0219483790
76180-696EA 60.89 USD
76180-696 76180-698 76180-700
γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥99%, white powder
γ-(N-Morpholino)propanesulphonic acid (MOPS)

MOPS is a zwitterionic amino acid which acts as one of the "Good" buffers. It is a structural analog to MES. However, MOPS is a more suitable biological buffer than MES because the pH range is closer to pH 7.4.
Store at Room Temperature (15-30 °C)


  • Molecular Biology Reagent Grade
  • Presentation: White Powder
  • pKa: 7.2 at 25 °C(Lit.)
  • Concentrations higher than 20 mM should not be used with mammalian cell cultures. ΔpKa/ΔT: -0.013 to -0.015(Lit.)
  • Soluble in water (to approximately 33% w/w - clear, colorless solution).
  • Sterilization of solutions should be done by filter sterilization through a 0.2 µm filter.

Formula: C₇H₁₅NO₄S
MW: 209.27 g/mol
Melting Pt: 277 °C (lit.)
Flash Pt: 110
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
Merck Index: 13,06288
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67e1af5961ad4d5fb4b8273ca2ca70a747b74367&vid=67e1af5961ad4d5fb4b8273ca2ca70a747b74367&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
UV/VIS: (1M aq soln) OD250 nm ≤ 0.04
UV/VIS: (1M aq soln) OD280 nm ≤ 0.04


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR