Pectinase, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0219897901 0219897910 0219897925 0219897980
76177-386EA 44.23 USD
76177-386 76177-390 76177-392 76177-394
Pectinase, MP Biomedicals
Enzymes
Pectinase is a macerating enzyme comprised of pectintranseliminase, polygalacturonase, pectinesterase, xylanase, hemicellulases and cellulases.
Pectinase is used in plant protoplast preparation to digest cell wall prior to organelle isolation.
Pectinase causes random hydrolysis of 1,4-alpha-D-galactosiduronic linkages in pectate and other galacturonans.
Store at +4 °C, desiccate.
  • Presentation: Beige Free Flowing Powder