X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥98%, white powder for molecular biology

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: BCIGX-GAL(2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

0219481190 0219481110 0219481180
76177-930EA 795.93 USD
76177-930 76177-926 76177-928
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥98%, white powder for molecular biology
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

Storage: Store at -20 °C. Store Desiccated. Store Under Nitrogen. Protect from light.
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside, commonly known as X−Gal, is a histochemical substrate for β−galactosidase.
5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside is used as indigogenic substrate for β-galactosidase, for detection of β-galactosidase-positive clones, and the identification of lac and bacterial colonies or phage plaques. It is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype. X−Gal is cleaved by β−galactosidase to yield an insoluble blue precipitate. X−Gal is particularly useful in molecular biology applications to detect the activity of β−galactosidase which is frequently used as a reporter gene. In cloning, X−Gal is used to detect insertion of foreign DNA into the lacZ region of plasmid DNA using α-complementation which is based on vectors such as the pUC and the M13mp series that carry a fragment of the β-galactosidase gene encoding an α-fragment of β-galactosidase.


  • Molecular Biology Reagent Grade
  • Optical Rotation: (c=1, DMF:H2O 1:1): -62.0 ° (Lit.)
  • Soluble in 50 mg/mL, DMF : MeOH (1:1). Soluble in DMF (2% w/v - clear, colorless solution).

Formula: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
Melting Pt: 230 °C (dec., lit.)
Storage Temperature: Freezer
MDL Number: MFCD00005666
CAS Number: 7240-90-6

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR