L-Methionine ≥99%, white crystalline powder cell culture reagent
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
(S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
0219470780
0219470791
0219470725
76177-868EA
110.91
USD
76177-868
76177-872
76177-914
L-Methionine ≥99%, white crystalline powder cell culture reagent
L-Methionine
Storage: Store at Room Temperature (15-30 °C).
Methionine is one of the common sulfur-containing amino acids. Methionine is a common methyl-group donor to various substrates, such as creatine, epinephrine, ergosterol, and choline.
- Cell Culture Reagent Grade
- Presentation: White Crystalline Powder
- pKa: 2.28, 9.21 (Lit.)
- Soluble at 2.5 g to 100 mL of water.
Formula:
C₅H₁₁NO₂S Melting Pt: 280…282 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00063097 CAS Number: 63-68-3 |
|
Specification Test Results
Identity Test | Passes |
Purity | ≥99% |
FTIR | Conforms to Standard |
pH (1% aq soln) | 5.6-6.1 |
Loss on Drying | ≤0.3% |
Optical Rotation (c=2, 6N HCl) | +22.4° to 24.7° |
Residue on Ignition | ≤0.4% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...