myo-Inositol, white powder cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol

0219468850 0219468880 0219468890 0219468891
76177-860EA 72.21 USD
76177-860 76177-862 76177-864 76177-866
myo-Inositol, white powder cell culture reagent
myo-Inositol

Storage: Store at Room Temperature (15-30 °C).
D-myo-Inositol is a component of membrane phospholipids, glycosylphophatidylinositol anchors that bind glycoproteins to cell membranes, and inositol phosphate second messengers. It is a growth factor for animals as well as microorganisms. Myo-Inositol is the most abundant form of polyols that serves as a structural element of secondary messengers in eukaryotic cells. Inositol or cyclohexane-1,2,3,4,5,6-hexol is a chemical compound, a sixfold alcohol (polyol) of cyclohexane. It exists in nine possible stereoisomers, of which the most prominent form, widely occurring in nature, is cis-1,2,3,5-trans-4,6-cyclohexanehexol, or myo-inositol (former name meso-inositol). Inositol is a carbohydrate, though not a classical sugar. It has a taste which has been assayed at half the sweetness of table sugar. Used as a lipotropic agent.
A component of membrane phospholipids, glycosyl­phosphatidyl­inositol anchors that bind glycoproteins to cell membranes, and inositol phosphate second messengers. In addition, inositol serves as an important component of the structural lipids phosphatidylinositol (PI) and its various phosphates, the phosphatidylinositol phosphate (PIP) lipids.


  • Cell Culture Reagent Grade
  • Vapor Density: 6.2 (vs air) (Lit.)
  • Presentation: White Powder

Formula: C₆H₁₂O₆
Melting Pt: 220…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS Number: 87-89-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e140ea6cc93139d8ecc7f04ce7092d5ec2053425&vid=4e22294e7081ea2e963b152510e149d0924e1350&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Solubility (10% aq.) Clear Solution
pH (10% aq.) Neutral to Litmus


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR