Benzyl alcohol, clear liquid ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: PhenylmethanolBenzenemethanolPhenylcarbinol

0219388290
76177-666EA 231.3 USD
76177-666
Benzyl alcohol, clear liquid ACS
Benzyl alcohol

Storage: Store at Room Temperature (15-30 °C)
Benzyl alcohol (BnOH) is a monoaromatic primary alcohol widely used as a solvent and an intermediate in cosmetic formulations, pharmaceutical and flavor/fragrance industries. It shows bacteriostatic property. Commercial method for its synthesis is by the alkaline hydrolysis of benzyl chloride. Conformational studies by ab initio calculations shows gauche cis conformation around the alcoholic group.
Benzyl alcohol may be used for the benzylation of toluene.


  • ACS Reagent Grade
  • Presentation: Clear Liquid
  • Auto Ignition: 436 °C (817 °F) (Lit.)
  • Vapor Density: 3.7 (vs air) (Lit.)
  • Vapor Pressure: 0.03 hPa at 20 °C ; 13.3 mmHg ( 100 °C), 3.75 mmHg ( 77 °C) (Lit.)
  • Refractive Index: n20/D 1.539 (Lit.)

Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205.4 °C (1013 hPa) (lit.)
Melting Pt: –15.3 °C (lit.)
Density: 1.0442 g/cm³ (22.5 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
https://us.vwr.com/stibo/web/std.lang.all/97/87/2299787.jpg

Order Now

Specification Test Results

Identity Test Passes
Color (APHA) ≤20
Residue after Ignition ≤0.005%
Acetophenone ≤0.02%
Benzaldehyde ≤0.01%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR