Weiler® Cable Twist Knot Wire Wheel, ORS Nasco
Supplier: Safety & Industrial Supplies
804-08565 804-08534 804-13266 804-13276 804-08615 804-13266P
300043-820EA 62.03 USD
300043-820 300045-448 300010-170 300010-171 300042-704 300045-100
Weiler® Cable Twist Knot Wire Wheel, ORS Nasco
Brushes
Maximum impact for extremely severe applications
Ideally suited for roughening, deburring, or cleaning applications. Brushes may be used with straight grinders, right angle grinders, and bench/pedestal grinders. Use to remove rust, paint, oxides, and scale.
Minimal flexibility.