Weiler® Narrow Face Crimped Wire Wheel, ORS Nasco
Supplier: Safety & Industrial Supplies
804-00184 804-01178 804-01675 804-01258 804-01155 804-01075 804-01135 804-01475 804-00144 804-01705 804-13081 804-01055 804-01065 804-01045 804-01898 804-01075P 804-01145 804-01165 804-01775 804-01805 804-00134 804-00154 804-00204
300010-120EA 70.7 USD
300010-120 300010-124 300010-125 300042-124 300044-102 300010-122 300044-098 300044-566 300045-774 300010-126 300010-167 300044-460 300044-462 300010-121 300043-086 300044-096 300044-100 300044-104 300044-108 300044-110 300045-770 300045-772 300045-776 300046-488
Weiler® Narrow Face Crimped Wire Wheel, ORS Nasco
Brushes
These brushes can be used with straight grinders, bench/pedastal grinders, and right-angle grinders.
  • Flexible brushing action
  • Can be mounted in multiples
  • Consistent performance
Brushes are designed for cleaning, light deburring, edge blending, roughening, and finishing applications. Use to remove rust, paint, oxides, and scale.