D(-)-Sorbitol ≥98%, white powder for molecular biology
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
(2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
May be used for washing spheroplasts and in isoelectric focusing to minimize endoosmotic flow in agarose gels. May be used to induce osmotic stress.
D-Sorbitol is a sugar alcohol that is commonly used as a sugar substitute. It occurs naturally and is also produced synthetically from glucose. The food industry uses D-sorbitol as an additive in the form of a sweetener, humectant, emulsifier, thickener, or dietary supplement. D-Sorbitol has also been found in cosmetics, paper, and pharmaceuticals. Naturally, D-sorbitol occurs widely in plants via photosynthesis, ranging from algae to higher order fruits of the family Rosaceae.
- Molecular Biology Reagent Grade
- Vapor Density: <1 (vs air)
- Vapor Pressure: <0.1 mmHg ( 25 °C)
- Presentation: White Powder
Formula:
HOH₂C(CH(OH))₄CH₂OH Boiling Pt: 105 °C Melting Pt: 95…105 °C Density: 1.47 g/ml (–5 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004708 CAS Number: 50-70-4 |
|
Specification Test Results
Identity Test | Passes |
Purity | ≥98% |
FTIR | Conforms to Standard |
Melting Point | 95 - 105°C |
Moisture | ≤1% |
Optical Rotation | -2° ± 2° (c = 2, water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...