Calcium chloride, free flowing white powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

0219381950 0219381980 0219381990 0219381991
76177-008EA 59.39 USD
76177-008 76177-010 76177-012 76177-014
Calcium chloride, free flowing white powder ACS
Calcium chloride

Calcium chloride (CaCl2) is inexpensive, easily available calcium salt with a wide range of applications. It is a key component in deicers as it melts snow and ice. Reports suggest that CaCl2 helps in retaining the firmness of fresh cut fruits and vegetables during the storage. Calcium chloride may be used in the following processes: • Modification of natural zeolite to increase its ability of fluoride removal. • Synthesis of calcium sulfate whiskers utilized in making polymer reinforcing composite materials. • Synthesis of calcium chloride-anodized aluminum composites, a solid sorbent. • As an extractant in the soil analysis process. • As a catalyst in the synthesis of aldols by reacting dimethylsilyl (DMS) enolates with aldehydes.


  • ACS Grade
  • Presentation: Free Flowing White Powder
  • Vapor Pressure: 0.01mmHg (20°C)

Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600 (lit.)
Melting Pt: 772 (lit.)
Density: 1.34 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
Merck Index: 14,01659
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a1366a04d95f48c10853d5240903029610e194b&vid=5a1366a04d95f48c10853d5240903029610e194b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Assay ≥96.0%
Titrable Base 0.006 meq/g
Magnesium & Alkali Salts ≤2.0%
Mesh Size No. 20 Sieve ≤10%
Mesh Size No. 200 Sieve ≤30%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR