1-Hexanol 98%, Reagent Grade

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane

H13303-1L H13303-2.5L
BJH13303-1LEA 75.68 USD
BJH13303-1L BJH13303-2.5L
1-Hexanol 98%, Reagent Grade
1-Hexanol
Formula: H₃C(CH₂)₄CH₂OH
Boiling Pt: 11.8 %(V)
Melting Pt: -52 °C
Density: 0.8 g/cm³ (20 °C)
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00002982
CAS Number: 111-27-3
EINECS: 203-852-3
UN: 2282
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a0793e56caf8b4e459c31d9f36b2eec3d50a4e4&vid=5117270d1d57dd4b9cca9ee3d787375106cc1216&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 97.5 %
APHA Max. 20
Vapor Density 4.5 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR