N,N-Dimethylformamide ≥99%, Reagent Grade
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF
D158550-1L
D158550-2.5L
BJD158550-5LEA
253.05
USD
BJD158550-5L
BJD158550-1L
BJD158550-2.5L
N,N-Dimethylformamide ≥99%, Reagent Grade
N,N-Dimethylformamide
- ≥95% for general laboratory use.
Beilstein Number: 605365
RTECS Number: LQ2100000
UNSPSC Code: 12191602
Formula:
HCON(CH₃)₂ Boiling Pt: 152-153 °C (1.013 hPa) Melting Pt: -61 °C Density: 0.94 g/cm³ (20 °C) Flash Pt: 57.5 °C (1.013.25 hPa) Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3,III |
|
Specification Test Results
Identity (IR) | complying |
Assay (GC) | min. 99.0 % |
Water (Karl Fischer) | max. 0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...