Hydrochloric acid 32 - 35%, TraceSELECT™ Ultra UltraTrace analysis, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

96208-500ML 96208-1L 96208-250ML
BJ96208-500MLEA 720.45 USD
BJ96208-500ML BJ96208-1L BJ96208-250ML
Hydrochloric acid 32 - 35%, TraceSELECT™ Ultra UltraTrace analysis, Fluka™
Hydrochloric acid

TraceSELECT™ Ultra and TraceSELECT


  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions


Beilstein Number: 1098214
RTECS Number: 1098214
UNSPSC Code: 12352301

Formula: HCl
MW: 36.46
Boiling Pt: 42 °C (1.013 hPa)
Melting Pt: –35 °C
Density: 1.190 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Assay 32.0 - 35.0 %
Density (D 20/4) 1.145 - 1.180
Silver (Ag) Max. 10 ppt
Aluminium (Al) Max. 50 ppt
Arsenic (As) Max. 100 ppt
Gold (Au) Max. 20 ppt
Barium (Ba) Max. 10 ppt
Beryllium (Be) Max. 10 ppt
Bismuth (Bi) Max. 10 ppt
Calcium (Ca) Max. 200 ppt
Cadmium (Cd) Max. 10 ppt
Cerium (Ce) Max. 10 ppt
Cobalt (Co) Max. 10 ppt
Chromium (Cr) Max. 100 ppt
Cesium (Cs) Max. 10 ppt
Copper (Cu) Max. 100 ppt
Dysprosium (Dy) Max. 10 ppt
Iron (Fe) Max. 200 ppt
Erbium (Er) Max. 10 ppt
Europium (Eu) Max. 10 ppt
Gallium (Ga) Max. 10 ppt
Gadolinium (Gd) Max. 10 ppt
Hafnium (Hf) Max. 10 ppt
Mercury (Hg) Max. 500 ppt
Holmium (Ho) Max. 10 ppt
Indium (In) Max. 10 ppt
Potassium (K) Max. 100 ppt
Lanthanum (La) Max. 10 ppt
Lithium (Li) Max. 10 ppt
Lutetium (Lu) Max. 10 ppt
Magnesium (Mg) Max. 50 ppt
Manganese (Mn) Max. 10 ppt
Molybdenum (Mo) Max. 50 ppt
Sodium (Na) Max. 100 ppt
Niobium (nb) Max. 10 ppt
Neodymium (nd) Max. 10 ppt
Nickel (Ni) Max. 50 ppt
Lead (Pb) Max. 10 ppt
Palladium (Pd) Max. 10 ppt
Praseodymium (Pr) Max. 10 ppt
Platinum (Pt) Max. 10 ppt
Rubidium (Rb) Max. 10 ppt
Rhenium (Re) Max. 10 ppt
Rhodium (Rh) Max. 10 ppt
Ruthenium (Ru) Max. 10 ppt
Antimony (Sb) Max. 100 ppt
Scandium (Sc) Max. 20 ppt
Selenium (Se) Max. 1000 ppt
Samarium (Sm) Max. 10 ppt
Tin (Sn) Max. 200 ppt
Strontium (Sr) Max. 10 ppt
Terbium (Tb) Max. 10 ppt
Tellurium (Te) Max. 10 ppt
Thorium (Th) Max. 10 ppt
Titanium (Ti) Max. 50 ppt
Thallium (Tl) Max. 10 ppt
Thulium (Tm) Max. 10 ppt
Uran (U) Max. 10 ppt
Vanadium (V) Max. 10 ppt
Tungsten (W) Max. 10 ppt
Yttrium (Y) Max. 10 ppt
Ytterbium (Yb) Max. 10 ppt
Zinc (Zn) Max. 100 ppt
Zirconium (Zr) Max. 10 ppt
Vapor Pressure 3.23 psi ( 21.1 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR