Sulfuric acid 95, TraceSELECT™ Ultra UltraTrace analysis, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
77239-1L
77239-250ML
77239-500ML
BJ77239-1LEA
1206.61
USD
BJ77239-1L
BJ77239-250ML
BJ77239-500ML
Sulfuric acid 95, TraceSELECT™ Ultra UltraTrace analysis, Fluka™
Sulfuric acid
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Formula:
H₂SO₄ MW: 98.08 Boiling Pt: 330 Melting Pt: 10.38 Density: 1.840 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8 II |
Specification Test Results
Assay | 94.5 - 97.0 % |
Silver (Ag) | Max. 0.01 ppb |
Aluminium (Al) | Max. 0.5 ppb |
Arsenic (As) | Max. 1 ppb |
Barium (Ba) | Max. 0.02 ppb |
Beryllium (Be) | Max. 0.01 ppb |
Bismuth (Bi) | Max. 0.01 ppb |
Calcium (Ca) | Max. 2 ppb |
Cadmium (Cd) | Max. 0.01 ppb |
Cerium (Ce) | Max. 0.01 ppb |
Cobalt (Co) | Max. 0.01 ppb |
Chromium (Cr) | Max. 0.05 ppb |
Cesium (Cs) | Max. 0.01 ppb |
Copper (Cu) | Max. 0.5 ppb |
Dysprosium (Dy) | Max. 0.01 ppb |
Erbium (Er) | Max. 0.01 ppb |
Europium (Eu) | Max. 0.01 ppb |
Iron (Fe) | Max. 0.5 ppb |
Gallium (Ga) | Max. 0.01 ppb |
Gadolinium (Gd) | Max. 0.01 ppb |
Hafnium (Hf) | Max. 0.01 ppb |
Mercury (Hg) | Max. 0.5 ppb |
Holmium (Ho) | Max. 0.01 ppb |
Indium (In) | Max. 0.01 ppb |
Potassium (K) | Max. 0.5 ppb |
Lanthanum (La) | Max. 0.01 ppb |
Lithium (Li) | Max. 0.02 ppb |
Lutetium (Lu) | Max. 0.01 ppb |
Magnesium (Mg) | Max. 0.2 ppb |
Manganese (Mn) | Max. 0.02 ppb |
Molybdenum (Mo) | Max. 0.05 ppb |
Sodium (Na) | Max. 1 ppb |
Niobium (nb) | Max. 0.01 ppb |
Neodymium (nd) | Max. 0.01 ppb |
Nickel (Ni) | Max. 0.1 ppb |
Lead (Pb) | Max. 0.05 ppb |
Palladium (Pd) | Max. 0.01 ppb |
Praseodymium (Pr) | Max. 0.01 ppb |
Platinum (Pt) | Max. 0.01 ppb |
Rubidium (Rb) | Max. 0.01 ppb |
Rhodium (Rh) | Max. 0.01 ppb |
Antimony (Sb) | Max. 0.05 ppb |
Scandium (Sc) | Max. 0.02 ppb |
Selenium (Se) | Max. 0.001 ppm |
Samarium (Sm) | Max. 0.01 ppb |
Tin (Sn) | Max. 0.5 ppb |
Strontium (Sr) | Max. 0.01 ppb |
Terbium (Tb) | Max. 0.01 ppb |
Tellurium (Te) | Max. 0.01 ppb |
Thorium (Th) | Max. 0.01 ppb |
Titanium (Ti) | Max. 0.1 ppb |
Thallium (Tl) | Max. 0.01 ppb |
Thulium (Tm) | Max. 0.01 ppb |
Uran (U) | Max. 0.01 ppb |
Vanadium (V) | Max. 0.01 ppb |
Tungsten (W) | Max. 0.01 ppb |
Yttrium (Y) | Max. 0.01 ppb |
Ytterbium (Yb) | Max. 0.01 ppb |
Zinc (Zn) | Max. 0.5 ppb |
Zirconium (Zr) | Max. 0.02 ppb |
Vapor Pressure | 1 mmHg ( 146 °C) |
Vapor Density | <0.3 (25 °C, vs air) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...