n-Pentane ≥99.0% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Pentane

76869-1L 76869-2.5L
BJ76869-1LEA 234.51 USD
BJ76869-1L BJ76869-2.5L
n-Pentane ≥99.0% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
n-Pentane
Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36…37 °C
Melting Pt: -130
Density: 0.631 g/cm³ (15 °C)
Flash Pt: <–35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Refractive Index n20/D 1.358 (lit.)
Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.357 - 1.360
Isopentane (GC) Max. 1 %
2-methylpentane (GC) Max. 1 %
High-boiling fraction Max. 1 ppm
Explosion Limit ~8.3 %
Autoignition Temperature 500 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR