Sodium chlorite ~80% (RT), Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Danger

71388-250G 71388-1KG
BJ71388-250GEA 110.9 USD
BJ71388-250G BJ71388-1KG
Sodium chlorite ~80% (RT), Puriss. p.a., Fluka™
Sodium chlorite
Formula: ClNaO₂
MW: 90.44 g/mol
Melting Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00003478
CAS Number: 7758-19-2
EINECS: 231-836-6
UN: 1496
ADR: 5.1,II

Order Now

Specification Test Results

Assay 77.0 - 83.0 %
Silver (Ag) Max. 5 ppm
Aluminium (Al) Max. 5 ppm
Barium (Ba) Max. 5 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 1000 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 10 ppm
Manganese (Mn) Max. 5 ppm
Molybdenum (Mo) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 5 ppm
Thallium (Tl) Max. 5 ppm
Zinc (Zn) Max. 5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR