Acetonitrile ≥99.5% (by GC)

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

60004-1L 60004-2.5L 60004-4X2.5L 60004-4X5L 60004-5L
BJ60004-1LEA 190.56 USD
BJ60004-1L BJ60004-2.5L BJ60004-4X2.5L BJ60004-4X5L BJ60004-5L
Acetonitrile ≥99.5% (by GC)
Acetonitrile

We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:


  • Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
  • Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.


Beilstein Number: 741857
RTECS Number: AL7700000
UNSPSC Code: 12191502

Formula: H₃CCN
Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: –46 °C
Density: 0.7857 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Refractive Index n20/D 1.344 (lit.)
Assay (GC) Min. 99.5 %
Water (Karl Fischer) Max. 0.1 %
Vapor Density 1.41 (vs air)
Vapor Pressure 72.8 mmHg ( 20 °C)
Explosion Limit 16 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR