1-Dodecanol ≥98.0% ACS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
1-Dodecyl alcohol
443816-1KG
BJ443816-1KGEA
200.32
USD
BJ443816-1KG
1-Dodecanol ≥98.0% ACS, Riedel-de Haën™
1-Dodecanol
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Formula:
H₃C(CH₂)₁₀CH₂OH MW: 186.34 g/mol Boiling Pt: 260-262 °C Melting Pt: 22-26 °C Flash Pt: 121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004753 CAS Number: 112-53-8 EINECS: 203-982-0 |
Specification Test Results
Identity (IR) | complying |
Assay (GC) | Min. 98.0 % |
Water (Karl Fischer) | Max. 0.05 % |
APHA | Max. 10 |
Vapor Density | 7.4 (vs air) |
Explosion Limit | 4 % |
Autoignition Temperature | 500 °F |
Vapor Pressure | 0.1 mmHg ( 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...