Pyridine ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Azabenzene, Azine
360570-100ML
360570-1L
360570-2.5L
BJ360570-100MLEA
184
USD
BJ360570-100ML
BJ360570-1L
BJ360570-2.5L
BJ360570-500ML
Pyridine ≥99.0% ACS, Riedel-de Haën™
Pyridine
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Formula:
C₅H₅N Boiling Pt: 115 °C (1.013 hPa) Melting Pt: -42 °C Density: 0.98 g/cm³ (20 °C) Flash Pt: 12.4 %(V) Storage Temperature: Ambient |
MDL Number:
MFCD00011732 CAS Number: 110-86-1 EINECS: 203-809-9 UN: 1282 ADR: 3,II |
|
Specification Test Results
Identity (IR) | complying |
Assay (GC) | Min. 99.0 % |
Non-volatile matter | Max. 0.002 % |
Water (Karl Fischer) | Max. 0.1 % |
Ammonia (NH₃) | Max. 0.002 % |
Copper (Cu) | Max. 0.0005 % |
Chloride (Cl) | Max. 0.001 % |
Sulfate (SO₄) | Max. 0.001 % |
Solubility in water | complying |
KMnO₄ reducing substances | complying |
Appearance | complying |
Vapor Density | 2.72 (vs air) |
Vapor Pressure | 20 mmHg ( 25 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...