Methyl ethyl ketone ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
EK 500 Additive, Butan-2-one, Ethyl methyl ketone, 2-Butanone, Butanone, MEK
360473-1L
360473-6X1L
BJ360473-1LEA
115
USD
BJ360473-1L
BJ360473-6X1L
Methyl ethyl ketone ≥99.0% ACS, Riedel-de Haën™
Methyl ethyl ketone
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Beilstein Number: 741880
RTECS Number: EL6475000
UNSPSC Code: 12191602
Formula:
C₂H₅COCH₃ Boiling Pt: 79…81 °C Melting Pt: –87 °C Density: 0.805 g/cm³ (20 °C) Flash Pt: 11.5 %(V) Storage Temperature: Ambient |
MDL Number:
MFCD00011648 CAS Number: 78-93-3 EINECS: 201-159-0 UN: 1193 ADR: 3,II |
|
Specification Test Results
Refractive Index | n20/D 1.379 (lit.) |
Identity (IR) | complying |
Assay (GC) | Min. 99.0 % |
Non-volatile matter | Max. 0.0025 % |
Water (Karl Fischer) | Max. 0.20 % |
Titrable Acid | Max. 0.0005 meq/g |
APHA | Max. 15 |
Vapor Pressure | 71 mmHg ( 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...