Pyridine ≥99.5% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Azabenzene, Azine

33553-1L 33553-500ML
BJ33553-1LEA 395.86 USD
BJ33553-1L BJ33553-500ML
Pyridine ≥99.5% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, Riedel-de Haën™
Pyridine

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115
Melting Pt: -44
Density: 0.98 g/cm³ (20 °C)
Flash Pt: 12.4 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3 II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.5 %
Boiling range 114 - 116 °C
Explosion Limit 12.4 %
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.1 %
Ammonia (NH₃) Max. 0.001 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000003 %
Iron (Fe) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Chloride (Cl) Max. 0.0005 %
Sulfate (SO₄) Max. 0.0005 %
KMnO₄ reducing matter (as O) Max. 0.0005 %
Piperidine (GC) Max. 0.01 %
2-picoline (GC) Max. 0.2 %
Solubility in water complying
Refractive Index n20/D 1.509 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR