1-Propanol 99,5 GC_ASSAY_METHOD, Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
33538-1L
33538-2.5L
BJ33538-1LEA
169.19
USD
BJ33538-1L
BJ33538-2.5L
1-Propanol 99,5 GC_ASSAY_METHOD, Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
1-Propanol
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Beilstein Number: 1098242
Formula:
H₃CCH₂CH₂OH MW: 60.1 g/mol Boiling Pt: 97 Melting Pt: -127 Density: 0.803…0.804 g/cm³ (20 °C) Flash Pt: 10.8 %(V) Storage Temperature: Ambient |
MDL Number:
MFCD00002941 CAS Number: 71-23-8 EINECS: 200-746-9 UN: 1274 ADR: 3 II Merck Index: 13,07934 |
|
Specification Test Results
Assay (GC) | Min. 99.5 % |
Boiling range | 96 - 99 °C |
Density (D 20/20) | 0.802 - 0.806 |
Non-volatile matter | Max. 0.001 % |
Water (Karl Fischer) | Max. 0.05 % |
Free acid (as C₂H₅COOH) | Max. 0.001 % |
Aluminium (Al) | Max. 0.00005 % |
Boron (B) | Max. 0.000002 % |
Barium (Ba) | Max. 0.00001 % |
Calcium (Ca) | Max. 0.00005 % |
Cadmium (Cd) | Max. 0.000005 % |
Cobalt (Co) | Max. 0.000002 % |
Chromium (Cr) | Max. 0.000002 % |
Copper (Cu) | Max. 0.000002 % |
Iron (Fe) | Max. 0.00001 % |
Magnesium (Mg) | Max. 0.00001 % |
Manganese (Mn) | Max. 0.000002 % |
Nickel (Ni) | Max. 0.000002 % |
Lead (Pb) | Max. 0.00001 % |
Tin (Sn) | Max. 0.00001 % |
Zinc (Zn) | Max. 0.00001 % |
Acetone (GC) | Max. 0.01 % |
Ethanol (GC) | Max. 0.05 % |
Methanol (GC) | Max. 0.05 % |
2-propanol (GC) | Max. 0.05 % |
Propionaldehyde (GC) | Max. 0.01 % |
Autoignition Temperature | 700 °F |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...