Isobutanol ≥99% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 2-Methyl-1-propanol iso-Butyl alcohol iso-Butanol

33064-1L 33064-2.5L
BJ33064-1LEA 144.95 USD
BJ33064-1L BJ33064-2.5L
Isobutanol ≥99% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Isobutanol

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: (CH₃)₂CHCH₂OH
MW: 74.12 g/mol
Boiling Pt: 108 °C (1.013 hPa)
Melting Pt: -108 °C
Density: 0.802 g/cm³ (20 °C)
Flash Pt: 10.6 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/12/85/2301285.jpg

Order Now

Specification Test Results

Assay (GC) Min. 99 %
Boiling range 107 - 109 °C
Refractive Index n20/D 1.3950-1.3960
Boiling range 107-109 °C Min. 96 %
Density (D 20/4) 0.801 - 0.804
Refractive index (n 20/D) 1.3950 - 1.3960
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as C₃H₇COOH) Max. 0.004 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
1-Butanol (GC) Max. 0.1 %
2-Butanol (GC) Max. 0.05 %
n-Butyraldehyde (GC) Max. 0.005 %
iso-Butyraldehyde (GC) Max. 0.005 %
Butanone (GC) Max. 0.02 %
Peroxides (as H₂O₂) Max. 0.001 %
Reaction against H₂SO₄ complying
APHA Max. 10
Solubility in water complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR