Diethyl ether ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

32203-1L 32203-2.5L 32203-27L 32203-4X2.5L 32203-5L 32203-6X1L
BJ32203-1LEA 155.62 USD
BJ32203-1L BJ32203-2.5L BJ32203-27L BJ32203-4X2.5L BJ32203-5L BJ32203-6X1L
Diethyl ether ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Diethyl ether

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Beilstein Number: 1696894
RTECS Number: KI5775000
UNSPSC Code: 12191602

Formula: (CH₃CH₂)₂O
Boiling Pt: 34…35 °C (1.013 hPa)
Melting Pt: –116 °C (1.013 hPa)
Density: 0.71 g/cm³ (20 °C)
Flash Pt: –45 °C (1.013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (color) complying
Appearance (form) complying
Assay (GC) Min. 99.8 %
Boiling range 34 - 35 °C
Density (D 20/4) 0.713 - 0.714
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.03 %
Free acid (as CH₃COOH) Max. 0.0002 %
Titrable Acid Max. 0.0002 meq/g
Aluminium (Al) Max. 0.50 ppm
Boron (B) Max. 0.02 ppm
Barium (Ba) Max. 0.10 ppm
Calcium (Ca) Max. 0.50 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.05 ppm
Magnesium (Mg) Max. 0.10 ppm
Manganese (Mn) Max. 0.02 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.05 ppm
Tin (Sn) Max. 0.10 ppm
Zinc (Zn) Max. 0.05 ppm
Acetone (GC) Max. 0.005 %
Aldehydes complying
Carbonyl compounds (as CO) Max. 0.001 %
Carbonyl Compounds (as HCHO) Max. 0.001 %
Ethanol (GC) Max. 0.02 %
Methanol (GC) Max. 0.02 %
Peroxides (as H₂O₂) Max. 0.3 ppm
Reaction against H₂SO₄ complying
APHA Max. 10
Assay BHT (GC) 5 - 8 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR