Zinc sulfate concentrate for dilution to 1L, 0.1 M ZnSO4 volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
32047-1EA
32047-6X1EA
BJ32047-1EAEA
87.24
USD
BJ32047-1EA
BJ32047-6X1EA
Zinc sulfate concentrate for dilution to 1L, 0.1 M ZnSO4 volumetric solution, Fluka™
Zinc sulphate heptahydrate
For 1L standard solution
Formula:
ZnSO₄·7H₂O MW: 287.56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149894 CAS Number: 7446-20-0 |
Specification Test Results
Factor | 0.998 - 1.002 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...