1,10-Phenanthroline hydrochloride monohydrate 99.5-102% (dried basis) Reag. Ph. Eur. for spectrophotometry, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 1,10-Phenanthroline Chloride Monohydrate, 1,10-Phenanthrolinium chloride monohydrate, 1,10-Phenanthroline monohydrochloride monohydrate

31043-10G 31043-5G
BJ31043-10GEA 105 USD
BJ31043-10G BJ31043-500G BJ31043-5G
1,10-Phenanthroline hydrochloride monohydrate 99.5-102% (dried basis) Reag. Ph. Eur. for spectrophotometry, Fluka™
1,10-Phenanthroline hydrochloride monohydrate
Formula: C₁₂H₉ClN₂·1H₂O
MW: 234.68 g/mol
Melting Pt: 224…227 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150061
CAS Number: 18851-33-7
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Identity (IR) complying
Assay 99.5 - 102.0 %
Melting point Min. 224 °C
Loss on drying (105°C) Max. 8 %
Sulfated ash Max. 0.2 %
Sensitive according Fe(II) ions complying
Solubility (10 % in H₂O) complying
Turbidity of solution complying
Colour of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR