Copper(II) chloride dihydrate ≥99.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate

307483-100G 307483-500G
BJ307483-100GEA 101.6 USD
BJ307483-100G BJ307483-500G
Copper(II) chloride dihydrate ≥99.0% ACS, Fluka™
Copper (II) chloride dihydrate
Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Boiling Pt: 993 °C (anhydr.)
Melting Pt: 100 (–H₂O of crystall.)
Density: 2.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
UN: 2802
ADR: 8 III
Merck Index: 13,02660

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
pH (5 %, 20°C) 3.0 - 3.8
Insoluble in hydrochloric acid Max. 0.01 %
Calcium (Ca) Max. 50 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 100 ppm
Sodium (Na) Max. 200 ppm
Nickel (Ni) Max. 100 ppm
Nitrate (NO₃) Max. 0.015 %
Sulfate (SO₄) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR