Trifluoroacetic acid ≥99.0%, CHROMASOLV™ for HPLC, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
TFA
302031-1L
302031-100ML
BJ302031-1LEA
1559.78
USD
BJ302031-1L
BJ302031-100ML
Trifluoroacetic acid ≥99.0%, CHROMASOLV™ for HPLC, Fluka™
Trifluoroacetic acid
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Beilstein Number: 742035
Formula:
CF₃COOH Boiling Pt: 72 °C (1.013 hPa) Melting Pt: -15 °C Density: 1.48 g/cm³ (20 °C) Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004169 CAS Number: 76-05-1 UN: 2699 ADR: 8,I |
|
Specification Test Results
Assay (alkalimetric) | 99.0 - 101.0 % |
Assay (GC) | Min. 99.0 % |
Identity (IR) | complying |
Non-volatile matter | Max. 0.005 % |
Water (Karl Fischer) | Max. 0.05 % |
Absorbance at 320 nm | Max. 0.025 |
Absorbance at 300 nm | Max. 0.03 |
Absorbance at 290 nm | Max. 0.04 |
Absorbance 280 nm | Max. 0.05 |
Absorbance at 270 nm | Max. 0.10 |
Absorbance at 260 nm | Max. 0.9 |
APHA | Max. 20 |
Vapor Density | 3.9 (vs air) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...