Xylene (mixture of isomers) ≥98.5% (xylenes + ethylbenzene basis) ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Dimethylbenzene

247642-1L-CB 247642-2.5L 247642-20L
BJ247642-1L-CBEA 125.13 USD
BJ247642-1L-CB BJ247642-2.5L BJ247642-20L
Xylene (mixture of isomers) ≥98.5% (xylenes + ethylbenzene basis) ACS, Riedel-de Haën™
Xylene (mixture of isomers)
Formula: C₆H₄(CH₃)₂
Boiling Pt: 136 - 140 °C
Melting Pt: -34 °C
Density: 0.87 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
UN: 1307
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Xylene isomers and Ethylbenzol (GC) Min. 98.50 %
Ethyl benzene (GC) Max. 25 %
Water (Karl Fischer) Max. 0.05 %
Non-volatile matter Max. 0.002 %
S-compounds (as S) Max. 0.003 %
APHA Max. 10
Reaction against H₂SO₄ complying
Vapor Pressure 18 mmHg ( 37.7 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR