Methanol ≥99.7% (by GC), Puriss., meets analytical specification of Ph.Eur., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Wood alcohol, Methyl alcohol, Methyl hydroxide, Hydroxy methane, Carbinol, Methylol

24229-160KG 24229-2.5L 24229-4X2.5L
BJ24229-160KGEA 1931.83 USD
BJ24229-160KG BJ24229-20KG BJ24229-25L BJ24229-2.5L BJ24229-4X2.5L
Methanol ≥99.7% (by GC), Puriss., meets analytical specification of Ph.Eur., Riedel-de Haën™
Methanol

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.


Beilstein Number: 1098229

Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6
Melting Pt: -98
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.7 %
Boiling range 64 - 65 °C
Density (D 20/20) 0.791 - 0.793
Refractive index (n 20/D) 1.328 - 1.330
Non-volatile matter Max. 10 ppm
Water (Karl Fischer) Max. 0.10 %
Acidity or alkalinity complying
Free acid (as HCOOH) Max. 0.003 %
Free alkali (as NH₃) Max. 0.0003 %
Iron (Fe) Max. 0.0001 %
Identity (IR) complying
Chloride (Cl) Max. 0.0001 %
Sulfate (SO₄) Max. 0.0005 %
Reaction against H₂SO₄ complying
Reducing impurities complying
Aldehydes, Ketones (as CH₃CocH₃) Max. 0.003 %
Acetone (GC) Max. 0.001 %
Benzene (GC) Max. 2 ppm
UV-absorption complying
Impurity A (GC) complying
Related substances (GC) complying
Appearance complying
Residual solvents complying
Refractive Index n20/D 1.329 (lit.)
Explosion Limit 36 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR