Sodium hexanitritocobaltate(III) ACS, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Sodium hexanitritocobaltate(III)
233722-500G
233722-100G
BJ233722-100GEA
206.37
USD
BJ233722-100G
BJ233722-500G
Sodium hexanitritocobaltate(III) ACS, Fluka™
Sodium hexanitritocobaltate(III)
Formula:
CoN₆Na₃O₁₂ MW: 403.94 g/mol Melting Pt: 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003512 CAS Number: 13600-98-1 EINECS: 237-077-7 UN: 1479 ADR: 5.1,II |
Specification Test Results
Appearance of the substance | complying |
Insoluble in acetic acid | Max. 0.020 % |
Suitability for K determination | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...