n-Hexane ≥95%, Laboratory Reagent
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Hexyl hydride, Hexanen, Hexane
208752-25L
208752-1L
208752-2.5L
208752-4X2.5L
BJ208752-190LEA
4890.73
USD
BJ208752-190L
BJ208752-20L
BJ208752-25L
BJ208752-1L
BJ208752-2.5L
BJ208752-4X5L
BJ208752-5L
BJ2087524X2.5L
n-Hexane ≥95%, Laboratory Reagent
n-Hexane
- ≥95% for general laboratory use.
Beilstein Number: 1730733
RTECS Number: MN9275000
UNSPSC Code: 12191502
Formula:
H₃C(CH₂)₄CH₃ Boiling Pt: 66…69 °C(1.013 hPa) Melting Pt: –95 °C Density: 0.660…0.668 g/cm³ (20 °C) Flash Pt: –26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009520 CAS Number: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3,II |
|
Specification Test Results
Autoignition Temperature | 453 °F |
IR spectrum | complying |
Assay (GC) | Min. 95.0 % |
Density (D 20/20) | 0.658 - 0.662 |
Refractive index (n 20/D) | 1.373 - 1.377 |
Non-volatile matter | Max. 0.0015 % |
Water (Karl Fischer) | Max. 0.015 % |
Aromatic substances (as C₆H₆) | Max. 0.005 % |
Vapor Pressure | 5.2 psi ( 37.7 °C) |
Explosion Limit | 7.7 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...