Lithium perchlorate ≥95.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Cobaltous chloride hexahydrate Cobalt dichloride hexahydrate

205281-100G 205281-500G
BJ205281-100GEA 123.15 USD
BJ205281-100G BJ205281-500G
Lithium perchlorate ≥95.0% ACS, Fluka™
Lithium perchlorate
Formula: CoCl₂·6H₂O
MW: 106.39 g/mol
Melting Pt: 236 °C
Density: 2.420 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
UN: 1481
ADR: 5.1,II

Order Now

Specification Test Results

Assay Min. 95.0 %
pH (5 %, 25 °C) 6.0 - 7.5
Water insoluble matter Max. 0.005 %
Iron (Fe) Max. 5 ppm
Heavy metals (ICP) ACS Max. 5 ppm
Chloride (Cl) Max. 0.003 %
Sulfate (SO₄) Max. 0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR