2-Propanol ≥99.5% ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

190764-2.5L 190764-1L 190764-500ML 190764-6X1L 190764-6X500ML
BJ190764-2.5LEA 254 USD
BJ190764-2.5L BJ190764-1L BJ190764-500ML BJ190764-6X1L BJ1907646500ML
2-Propanol ≥99.5% ACS, Riedel-de Haën™
2-Propanol

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Beilstein Number: 635639
RTECS Number: NT8050000
UNSPSC Code: 12191601

Formula: (CH₃)₂CHOH
Boiling Pt: 77…83 °C (1.013 hPa)
Melting Pt: –90 °C
Density: 0.784…0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Explosion Limit 2 %
Assay (GC) Min. 99.50 %
IR spectrum complying
Solubility in water complying
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.2 %
Free acid, free alkali Max. 0.0001 meq/g
Carbonyl compounds (as Acetone) Max. 0.002 %
Carbonyl compounds (as propionaldehyde) Max. 0.002 %
APHA Max. 10
Autoignition Temperature 750 °F
Refractive Index n20/D 1.377 (lit.)
Vapor Pressure 33 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR