Acetone ≥99.5%, Laboratory Reagent

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

179973-25L 179973-1L 179973-4L 179973-4X4L 179973-6X1L
BJ179973-25LEA 624.09 USD
BJ179973-25L BJ179973-1L BJ179973-4L BJ179973-4X4L BJ179973-5L BJ179973-6X1L
Acetone ≥99.5%, Laboratory Reagent
Acetone

  • ≥95% for general laboratory use.


Beilstein Number: 635680
RTECS Number: AL3150000
UNSPSC Code: 12191602

Formula: CH₃COCH₃
Boiling Pt: 55…57 °C (1.013 hPa)
Melting Pt: –95 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

IR spectrum complying
Assay (GC) Min. 99.50 %
Refractive index (n 20/D) 1.357 - 1.361
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.5 %
Explosion Limit 13.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR