Methanol ≥99.6%, Laboratory Reagent
Supplier: Honeywell Research Chemicals
|
Danger
|
179957-2.5L
179957-4X2.5L
BJ179957-2.5LEA
122.01
USD
BJ179957-2.5L
BJ1799574X2.5L
Methanol ≥99.6%, Laboratory Reagent
Methanol
- ≥95% for general laboratory use.
Beilstein Number: 1098229
Formula:
CH₃OH Boiling Pt: 64…65 °C (1.013 hPa) Melting Pt: –98 °C Density: 0.79 g/cm³ (20 °C) Flash Pt: 11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3,II |
|
Specification Test Results
Explosion Limit | 36 % |
Identity (IR) | complying |
Assay (GC) | Min. 99.6 % |
Non-volatile matter | Max. 0.01 % |
Water (Karl Fischer) | Max. 0.1 % |
Free acid (as HCOOH) | Max. 0.03 % |
Free alkali (as NH₃) | Max. 0.01 % |
Vapor Pressure | 410 mmHg ( 50 °C) |
Autoignition Temperature | 725 °F |
Vapor Density | 1.11 (vs air) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...