Tetrachloroethylene ≥99.5% stabilized
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Tetrachloroethene
, PCE
, Ethylene tetrachloride
, Perchloroethylene
16211-6X1L
16211-1L
16211-2.5L
BJ16211-6X1LCS
565.16
USD
BJ16211-6X1L
BJ16211-2.5L
BJ16211-1L
Tetrachloroethylene ≥99.5% stabilized
Tetrachloroethylene
We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
- Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
- Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Beilstein Number: 1361721
RTECS Number: KX3850000
UNSPSC Code: 12190000
Formula:
C₂Cl₄ MW: 165.83 g/mol Boiling Pt: 120-122 °C (1.013 hPa) Melting Pt: -22 °C Density: 1.622…1.624 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000834 CAS Number: 127-18-4 EINECS: 204-825-9 UN: 1897 ADR: 6.1,III |
Specification Test Results
Vapor Density | 5.83 (vs air) |
Contains | stabilizer |
Assay | Min. 99.5 % |
Non-volatile matter | Max. 0.001 % |
Water (Karl Fischer) | Max. 0.01 % |
Free acid (as HCl) | Max. 0.001 % |
Refractive Index | n20/D 1.505 (lit.) |
Vapor Pressure | 19 mmHg ( 25 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...