n-Pentane 98%, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger

158941-2.5L 158941-1L
BJ158941-2.5LEA 174.14 USD
BJ158941-2.5L BJ158941-1L
n-Pentane 98%, Reagent Grade
n-Pentane

  • ≥95% for general laboratory use.


Beilstein Number: 969132
RTECS Number: RZ9450000
UNSPSC Code: 12191502

Formula: H₃C(CH₂)₃CH₃
Boiling Pt: 36…37 °C
Melting Pt: –131 °C
Density: 0.631 g/cm³ (15 °C)
Flash Pt: <–35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Density 2.48 (vs air)
Assay (GC) Min. 97.5 %
Refractive index (n 20/D) 1.356 - 1.360
IR spectrum complying
Refractive Index n20/D 1.358 (lit.)
Explosion Limit ~8.3 %
Autoignition Temperature 500 °F
Vapor Pressure 8.43 psi ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR