Benzyl alcohol ≥99.0% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Benzenemethanol, Phenylmethanol, Phenylcarbinol
13160-500ML
13160-2.5L
BJ13160-500MLEA
285.7
USD
BJ13160-500ML
BJ13160-2.5L
Benzyl alcohol ≥99.0% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Benzyl alcohol
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Beilstein Number: 878307
RTECS Number: DN3150000
UNSPSC Code: 12191503
Formula:
C₆H₅CH₂OH Boiling Pt: 205.3 °C (1.013 hPa) Melting Pt: -15.4 °C Density: 1.045 g/cm³ (20 °C) Flash Pt: 100.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004599 CAS Number: 100-51-6 EINECS: 202-859-9 |
|
Specification Test Results
Assay (GC) | Min. 99.0 % |
Refractive index (n 20/D) | 1.540 - 1.542 |
Non-volatile matter | Max. 0.005 % |
Sulfated ash | Max. 0.005 % |
Acetophenone (GC) | Max. 0.02 % |
Benzaldehyde (GC) | Max. 0.01 % |
Free acid (as C₆H₅COOH) | Max. 0.03 % |
Appearance of the solution | complying |
Aluminium (Al) | Max. 0.5 ppm |
Barium (Ba) | Max. 0.1 ppm |
Bismuth (Bi) | Max. 0.1 ppm |
Calcium (Ca) | Max. 0.5 ppm |
Cadmium (Cd) | Max. 0.05 ppm |
Cobalt (Co) | Max. 0.02 ppm |
Chromium (Cr) | Max. 0.02 ppm |
Copper (Cu) | Max. 0.02 ppm |
Iron (Fe) | Max. 0.5 ppm |
Potassium (K) | Max. 0.5 ppm |
Lithium (Li) | Max. 0.1 ppm |
Magnesium (Mg) | Max. 0.1 ppm |
Manganese (Mn) | Max. 0.02 ppm |
Molybdenum (Mo) | Max. 0.1 ppm |
Sodium (Na) | Max. 2 ppm |
Nickel (Ni) | Max. 0.02 ppm |
Lead (Pb) | Max. 0.1 ppm |
Strontium (Sr) | Max. 0.1 ppm |
Zinc (Zn) | Max. 0.1 ppm |
APHA | Max. 20 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...