2-Ethoxyethanol 99%, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Ethyl glycol, Ethylene glycol monoethyl ether, Cellosolve

128082-1L 128082-2.5L 128082-25L
BJ128082-1LEA 83.67 USD
BJ128082-1L BJ128082-2.5L BJ128082-25L
2-Ethoxyethanol 99%, Reagent Grade
2-Ethoxyethanol
Formula: C₂H₅OCH₂CH₂OH
MW: 90.12 g/mol
Boiling Pt: 135
Melting Pt: -70
Density: 0.93 g/cm³ (20 °C)
Flash Pt: 40 °C (1.013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00002869
CAS Number: 110-80-5
EINECS: 203-804-1
UN: 1171
ADR: 3 III
Merck Index: 13,03786
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=461a4daf8709ebd6b4516d73b4b32eee33cd1bca&vid=625adefb8d1e2126363931c57a51d3e1df419926&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 98.5 %
Water (Karl Fischer) Max. 0.10 %
APHA Max. 10
Autoignition Temperature 460 °F
Refractive Index n20/D 1.407 (lit.)
Vapor Pressure 3.8 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR