Heptane (mixture of isomers) ≥99.8%, TraceSELECT™ for inorganic trace analysis, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: DipropylmethaneEptaniHeptanHeptaneHeptanenHeptyl hydrideSkellysolve c

01379-1L
BJ01379-1LEA 253.26 USD
BJ01379-1L
Heptane (mixture of isomers) ≥99.8%, TraceSELECT™ for inorganic trace analysis, Riedel-de Haën™
Heptane (mixture of isomers)

High purity solvents for speciation analysis by LC-ICP-MS. Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance. The blank values for metal traces in these solvents are in the ppb range or lower.

Formula: H₃C(CH₂)₅CH₃
MW: 100.2 g/mol
Boiling Pt: 98.2-98.4 °C (1.000 hPa)
Melting Pt: -90.5 °C
Density: 0.69 g/cm³ (15 °C)
Flash Pt: -4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/64/15/736415.jpg

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Water (Karl Fischer) Max. 0.005 %
Silver (Ag) Max. 0.5 ppb
Aluminium (Al) Max. 1 ppb
Arsenic (As) Max. 1 ppb
Gold (Au) Max. 0.5 ppb
Barium (Ba) Max. 0.5 ppb
Beryllium (Be) Max. 0.5 ppb
Bismuth (Bi) Max. 0.5 ppb
Calcium (Ca) Max. 20 ppb
Cadmium (Cd) Max. 0.5 ppb
Cerium (Ce) Max. 0.5 ppb
Cobalt (Co) Max. 0.5 ppb
Chromium (Cr) Max. 1 ppb
Cesium (Cs) Max. 0.5 ppb
Copper (Cu) Max. 1 ppb
Iron (Fe) Max. 1 ppb
Gallium (Ga) Max. 0.5 ppb
Germanium (Ge) Max. 0.5 ppb
Mercury (Hg) Max. 1 ppb
Indium (In) Max. 0.5 ppb
Iridium (Ir) Max. 0.5 ppb
Potassium (K) Max. 10 ppb
Lithium (Li) Max. 0.5 ppb
Magnesium (Mg) Max. 1 ppb
Manganese (Mn) Max. 0.5 ppb
Molybdenum (Mo) Max. 0.5 ppb
Sodium (Na) Max. 100 ppb
Nickel (Ni) Max. 1 ppb
Lead (Pb) Max. 0.5 ppb
Palladium (Pd) Max. 0.5 ppb
Platinum (Pt) Max. 0.5 ppb
Rubidium (Rb) Max. 0.5 ppb
Rhodium (Rh) Max. 0.5 ppb
Ruthenium (Ru) Max. 0.5 ppb
Antimony (Sb) Max. 0.5 ppb
Selenium (Se) Max. 1 ppb
Tin (Sn) Max. 0.5 ppb
Strontium (Sr) Max. 0.5 ppb
Titanium (Ti) Max. 1 ppb
Thallium (Tl) Max. 0.5 ppb
Vanadium (V) Max. 0.5 ppb
Zinc (Zn) Max. 1 ppb
Zirconium (Zr) Max. 0.5 ppb
Refractive Index n20/D 1.387 (lit.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR