Weiler® Vortec Pro® Knot Wire Cup Brush, ORS Nasco
Supplier: Safety & Industrial Supplies
804-36038 804-36039 804-36045 804-36041 804-36243 804-36244 804-36070 804-36044 804-36043 804-36238 804-36239 804-36245 804-36066
300085-462CS 162.54 USD
300085-462 300085-464 300085-474 300085-466 300085-468 300085-558 300085-560 300085-506 300085-472 300085-470 300085-550 300085-552 300085-554 300085-562 300086-384
Weiler® Vortec Pro® Knot Wire Cup Brush, ORS Nasco
Brushes
Applications include cleaning prior to welding, deburring, roughening, rust and paint removal, weld scale and corrosion removal.
  • Provides fast, efficient brushing action
  • For use on large, flat surfaces