Weiler® Vortec Pro® Crimped Wire Cup Brush, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-36236 804-36036 804-36033 804-36232 804-36061 804-36037 804-36234 804-36032 804-36068 804-36035 804-36034 804-36031 804-36261 804-36231
300086-374PK 56.12 USD
300086-374 300085-542 300085-454 300085-586 300085-456 300085-448 300085-536 300085-546 300085-458 300085-538 300085-504 300085-460 300085-450 300085-452
Weiler® Vortec Pro® Crimped Wire Cup Brush, ORS Nasco
Brushes
Cost-effective performance in general-purpose cleaning applications.
  • Provides fast, efficient, and flexible brushing action
  • For use on large, flat surfaces
Applications include cleaning prior to welding, light deburring, roughening, removal of weld scale, corrosion, paint, coatings, rust, and other foreign material from surfaces of bores and passages, and slag from the edges of plastic.