StatZap™ ESD-Eliminator, MicroCare
Supplier: MicroCare
MCC-ZAP
76020-260CS 237.45 USD
76020-260
StatZap™ ESD-Eliminator, MicroCare
Sprays
This is a handy dispensing tool designed to fit on to MicroCare™ aerosol cans and dissapate the static charge created by all high-pressure aerosol spray products, protecting delicate PCBs.
  • A handy dispensing tool for aerosol cans
  • Dissapates the ESD charge created by all aerosol sprays
  • Continuously drains static charges to less than 50 volts
  • Does not affect or impair solvent performance
  • Fits most MicroCare™ cans
The StatZap™ ESD Eliminator is a handy dispensing tool designed to dissapate the ESD charge created by all aerosol products. The StatZAP™ ESD Eliminator continuously drains static charges to less than 50 volts so the static can never reach levels dangerous to components and PCBs. Each StatZAP™ fits most MicroCare™ cans and many of those from other vendors, as well. By maintaining direct contact with the StatZAP™ trigger, the user becomes part of the grounding circuit and ESD charges are carried away to ground. Each StatZAP™ is made from stainless steel so it is durable and long-lasting. The tapered shape of the CoronaTip™ on the StatZAP™ generates an static-killing ionized triboelectric field that radiates as much as ten inches from the tip. This neutralizes ESD charges within that field. The ShockShield™ safety feature protects technicians from contact with energized electrical circuits. If static control is important, then the StatZap™ is vital. (Note: The operator must be grounded during use.) The StatZAP™ does neither improve nor impair solvent performance.
Caution: The operator must be grounded during use.