Weiler® Banded Crimped Wire End Brushes, ORS Nasco
Supplier: ORS Nasco
804-11113 804-11114 804-11112 804-11103 804-11110
300047-718EA 37.84 USD
300047-718 300048-446 300046-754 300047-716 300047-714
Weiler® Banded Crimped Wire End Brushes, ORS Nasco
Brushes
Crimped brushes are ideal for severe applications to remove rust, paint, oxides, and scale.
  • Banded, solid-end prevents flaring
  • Banded brushes provide shorter trim length for more aggressive brushing action
  • Use for parts that could be damaged by the impact of a knot brush
Use with power drills, drill presses, die grinders, and right-angle air die grinders.