Weiler® Stem-Mounted Circular Flared End Brush, ORS Nasco
Supplier: Safety & Industrial Supplies
804-10151 804-10050 804-10039 804-10041 804-10150 804-10037 804-10038 804-10046 804-10076
300045-786EA 166.73 USD
300045-786 300045-780 300091-884 300045-778 300045-784 300047-706 300047-708 300047-710 300048-744
Weiler® Stem-Mounted Circular Flared End Brush, ORS Nasco
Brushes
Flared end brushes are ideal for severe applications such as deburring, cleaning, and finishing.
  • Accommodates the natural working angle
  • Designed for a broad range of surfaces and contours
Use with die grinders and right-angle air die grinders.