Weiler® Hollow-End Knot Wire End Brush, ORS Nasco
Supplier: Safety & Industrial Supplies
804-10025 804-10027 804-10142 804-10031 804-10217 804-10029 804-10028P 804-10032 804-10026 804-10028 804-10141 804-10030 804-10208
300010-156EA 30.6 USD
300010-156 300010-158 300046-300 300046-684 300085-294 300087-842 300046-682 300046-686 300010-157 300010-159 300046-298 300047-446 300091-886
Weiler® Hollow-End Knot Wire End Brush, ORS Nasco
Brushes
Knot brushes are ideal for severe applications to remove rust, paint, oxides, and scale.
  • Hollow-end wire brush
  • Rugged knot-type construction provides aggressive, high-impact action for severe applications
  • Ideal for cleaning in recessed areas
Use with power drills, drill presses, die grinders, and right-angle air die grinders.