3M™ Betapure™ NT-T Series Filter Cartridge
Supplier: 3M Healthcare
Betapure™
7010303084 7000125467 7000125463 7010313722 7100029916 7100066074 7010413393 7010415277 7010313973 7100027478 7010380076 7010379412 7010379238 7000125602 7010338641 7000125468 7000050258 7010338646 7100066073 7010410573 7010383109 7000125352 7000050257 7000125464 7010301116 7010338021 7010411042 7010381358 7010413098 7010291476 7010303348 7000125466 7100169690 7010338662 7100006642 7010379462 7100167395 7100020594 7010381770 7000050760 7000050649 7010415614 7010338018 7010339238 7010338048 7000050504 7010379417 7010381736 7000125351 7000050501 7100041891 7010314823 7010338202 7000050253 7000050254 7010413500 7100118766 7000029358 7100114489 7100006317 7000144810 7010412806 7000125603 7010339251 7000050503 7010380271 7010314653 7000050500 7100129122 7010301112 7010314790 7000050530 7010379407 7100169696 7000125465 7000125350 7010413519 7010379229 7010409449 7010338047 7100059114 7100066559 7000029263 7010301111 7010409883 7010413398 7010301291 7010415724 7100064478 7010379234 7010379460 7000050270 7100010275 7000050761 7010339339 7010314637 7010301297 7000125462 7010379461 7010338020 7010412828 7010314799 7000125620 7010382988 7010411178 7000029189 7010381151 7000125354 7000125514 7000125611 7100057729
76032-643CS 1956.3 USD
76032-643 76033-849 76032-641 76033-851 76033-853 76033-855 76033-857 76032-633 76032-639 76032-637 76033-859 76032-631 76033-863 76033-865 76033-901 76032-629 76033-861 76032-701 76032-667 76032-665 76032-705 76033-827 76033-829 76032-661 76033-675 76033-831 76033-677 76033-679 76031-585 76031-581 76033-671 76033-673 76032-657 76032-655 76032-659 76033-837 76033-841 76033-843 76033-845 76033-847 76033-681 76031-573 76032-689 76032-721 76032-687 76028-587 76032-725 76028-585 76032-681 76033-925 76032-685 76032-683 76033-775 76033-777 76033-897 76033-779 76033-899 76033-813 76032-719 76028-579 76033-771 76033-891 76033-893 76032-679 76028-595 76032-711 76028-593 76032-677 76032-713 76032-671 76033-815 76032-675 76032-673 76033-785 76033-821 76033-787 76033-825 76031-597 76032-709 76032-707 76033-660 76033-781 76031-595 76033-783 76033-662 76033-905 76033-753 76033-873 76033-875 76033-911 76033-877 76033-913 76033-879 76033-871 76032-699 76032-731 76028-577 76032-693 76033-917 76032-697 76032-695 76033-763 76033-887 76033-767 76033-921 76033-923 76033-769 76032-729 76033-881 76033-883 76033-761
3M™ Betapure™ NT-T Series Filter Cartridge
Filters Filter Cartridges
Betapure™ NT–T series filter cartridge is designed to deliver high performance and long service life along with direct cost savings by reducing the number of filters used. The resulting reduction in filter change–out frequency will decrease direct labor and filter disposal costs. Polypropylene media allows for broad chemical and temperature compatibility.
  • Absolute–rated filter provides consistent filtration quality
  • Polypropylene filter media allows for broad chemical and temperature compatibility
  • Ratings from 0.5 to 70 µm suit a wide range of applications
  • Exhibits superior particle retention under increasing differential pressure
Construction combines a polypropylene media with fluid distribution netting to form multiple layers, where critically positioned media flow channels allow greater movement of fluid from layer to layer. Filter cartridges range from 0.5 to 70 µm with various cartridge lengths from ⁹/₃₄" to 40". We combine three distinct media sections, made from multiple media/netting layers, to form the filter cartridge.
We utilize advanced design and construction for our Betapure™ NT–T Series filters to achieve a high level of filtration consistency. Combined with an exceptionally long service life, the Betapure NT–T Series filter's consistent performance gives you predictable results throughout the filters' usable life. 100% polypropylene construction of filter media, netting, core, and end connectors affords extensive chemical and thermal compatibility and no media migration. FDA CFR–21 compliant materials of construction.
Applications: Chemical and Hydrocarbon Processing, Acids, Bleach (sodium hypo–chlorite), Polyethylene and polypropylene manufacture, Amine sweetening and water flood, Food and Beverage, Bottled water particulate and turbidity reduction, Reverse osmosis membrane and spray nozzle protection, Diatomaceous earth or carbon fine trap and beverage blending, Rinsing and wash water, Fine Chemical and Electronics, Pre–RO filtration of high silt density index incoming water, Copper sulfate plating bath filtration in printed circuit board construction and color screen filtration for CRT production, Film and paper coatings, photographic film, Lens coatings and can coatings, High quality paints and ink, Machine tool lubrication, Chemicals, detergents, and waste water, Textiles, plating baths, pulp and paper, Process water and ground water remediation.